Viabank™ Bacterial Storage Beads
Toimittaja: MEDICAL WIRE EQUIPMENT
MWVIB MWVIG MWVIY MWVIR MWVIM
MEDWMWVIBEA 306 EUR
MEDWMWVIB MEDWMWVIG MEDWMWVIY MEDWMWVIR MEDWMWVIM
Viabank™ Bacterial Storage Beads
Pikatestit patogeenien ja allergeenien osoittamiseen Mikrobitestit
Viabank™ bacterial storage beads for long term storage of microorganisms at low temperatures.
  • Twenty glass beads per vial
  • Coloured caps match colour of beads
  • Available in multi coloured packs of red, yellow, green and blue or singular packs of each colour
  • Individual beads can be removed while frozen at any time for establishing fresh cultures