Foetal bovine serum, South American origin, Corning®
Toimittaja: Corning
35-079-CV
MDTC35-079-CVEA 410.27 EUR
MDTC35-079-CV
Foetal bovine serum, South American origin, Corning®
Seerumit
Foetal bovine serum sourced in South America and manufactured at facilities approved by the appropriate local authorities.
  • Triple sterile filtered (0,1 μm)
  • Low endotoxin (<30 EU/ml)
  • Low haemoglobin (<30 mg/dl)
Corning® sera products are analysed for quality and performance. Testing includes pH, osmolality, growth promotion, sterility (current USP method), mycoplasma, total protein, endotoxin and haemogloblin levels, and viral contamination. A full biochemical profile is available for each lot upon request.