Cell culture growth supplement, Trace Elements A, Corning®
Toimittaja: Corning
25-021-CI
MDTC25-021-CIEA 59.78 EUR
MDTC25-021-CI
Cell culture growth supplement, Trace Elements A, Corning®
Soluviljelyyn suplementit ja lisäaineet
Trace Elements A provides copper, zinc, iron, and selenium
The combination of Trace Elements A contains the trace elements of MCDB 104 and of MCDB 301.
Selenium aids in the detoxification of free radicals as a cofactor for GSH synthetase, while iron, copper, and zinc may be bound by serum protein.