Weighing dishes, aluminium
Toimittaja: MARIENFELD
6604900
MARI6604900EA 54.5 EUR
MARI6604900
Weighing dishes, aluminium
Punnitusastiat Punnitusastiat
Dishes made of aluminium.
  • Flat bottom ensures good standing on the worktop and keeps the samples securely in place
  • Manufactured free of grease or oil
  • Good chemical and temperature resistance
Suitable for moisture analysis in infra-red dryer.