Dishes, crystallising
Toimittaja: MARIENFELD
4210403 4210409 4210411 4210413 4210500 4210507 4210514
MARI4210403EA 48.8 EUR
MARI4210403 MARI4210409 MARI4210411 MARI4210413 MARI4210500 MARI4210507 MARI4210514
Dishes, crystallising
Haihdutusmaljat/Tarjottimet/Altaat Kiteytysmaljat
Crystallising dishes, made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Cylindrical shape with fused rim and flat bottom.