NANOCOLOR® Photometric Tube Test Kits for Phosphates (Ortho) and Phosphates (Total)

Toimittaja: MACHEREY-NAGEL

985081 985076 985079 985055 985095 985080
MANA985081EA 192 EUR
MANA985081 MANA985076 MANA985079 MANA985055 MANA985095 MANA985080
NANOCOLOR® Photometric Tube Test Kits for Phosphates (Ortho) and Phosphates (Total)
Ympäristöanalytiikan pikatestit

Photometric test kits for the determination of phosphates (ortho); phosphates (total) in water samples.


  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting


The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.


Sea water analysis: Yes

Remark: Measuring range on NANOCOLOR VIS II

Hazardous material: Yes

Sludge reagent set: No

Digestion set: No

NanOx Metal: Yes

NanOx N: No

Digestion temperature: 100 °C;120 °C



Toimitustiedot:

Rugged box with 20 test tubes. Sufficient for 20 tests.


Order Now

TEKNISET TIEDOT

Tyyppi MANA985081 MANA985076 MANA985079 MANA985055 MANA985095 MANA985080
Environmentally Preferable
Alsuta NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests
Säilytyslämpötila 15...25 °C 15...25 °C 15...25 °C 15...25 °C 15...25 °C 15...25 °C
Säilyvyys (kuukautta) 12 12 36 18 12 12
Paino 455 g (Including packaging) 364,5 g (Including packaging) 384,1 g (Including packaging) 467,5 g (Including packaging) 378,3 g (Including packaging) 433 g (Including packaging)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR