NANOCOLOR® Photometric Tube Tests Kits for sulphate
Toimittaja: MACHEREY-NAGEL
985062 985063 985060
MANA985062EA 165 EUR
MANA985062 MANA985063 MANA985060
NANOCOLOR® Photometric Tube Tests Kits for sulphate
Ympäristöanalytiikan pikatestit
Photometric test kits for the determination of sulphate in water samples.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (OD) tubes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Remark: Measuring range on NANOCOLOR VIS II
Toimitustiedot: Supplied in rugged box with 20 test tubes.