pH Papers, without color scale
Toimittaja: MACHEREY-NAGEL
90705 90711 90713 91106 91107 91108 91116 91117 91118 91126 91127 91128 90712 90701 90703
MANA90705EA 48.5 EUR
MANA90705 MANA90711 MANA90713 MANA91106 MANA91107 MANA91108 MANA91116 MANA91117 MANA91118 MANA91126 MANA91127 MANA91128 MANA90712 MANA90701 MANA90703
pH Papers, without color scale
Testipaperit, pH-liuskat pH-testiliuskat
These products are simple, completely impregnated indicator papers, which do not allow exact pH determinations, but pH adjustments during special chemical procedures. For this reason, no colour scale is provided with these papers.
  • Long term stable and protected against external influences due to plastic packaging
  • No accessories needed - always ready to use
  • Quick and easy determination of the pH