Test strips, swimming pool test
Toimittaja: MACHEREY-NAGEL
90752 90759
MANA90752EA 50.5 EUR
MANA90752 MANA90759
Test strips, swimming pool test
Ympäristöanalytiikan pikatestit
These strips are used for quick and easy determination of water qualities in swimming pool. The test provides valuable results about the water hardness, the total alkalinity, free and total chlorine and pH.
  • Easy dip-and-read procedure
  • Multi-stage colour scale
  • Provides reliable results within 30 seconds
  • No accessories necessary for evaluation
  • Desiccant in the stopper of tube container for optimal protection of the strips against humidity