Selluloosa MN 301 TLC-analyyseihin

Toimittaja: MACHEREY-NAGEL

816250.1 816250.5
MANA816250.1EA 222 EUR
MANA816250.1 MANA816250.5
Selluloosa MN 301 TLC-analyyseihin
Selluloosa

Cellulose adsorbent for thin-layer chromatography.


  • Native fibrous cellulose
  • Standard grade

Kaava: (C₁₂H₂₂O₁₁)n
Sulamispiste: 260…270 °C
Tiheys: 1,27…1.6 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081512
CAS-numero: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Order Now

Testitulosten erittely

Fibre length (95 %) 2 – 20 μm
Average degree of polymerization 400 – 500
Specific surface acc. to Blaine 15000 cm²/g
Fe ≤20 ppm
Cu ≤6 ppm
P ≤7 ppm
CH₂Cl₂ extract ≤0,25 %
Residue on ignition at 850 °C ≤1500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR