Flash chomatography cartridges, empty colums, CHROMABOND® Flash DL and connecting kits
Toimittaja: MACHEREY-NAGEL
732980 732981 732982 732983 732984 732985 732986 732987
MANA732980EA 345 EUR
MANA732980 MANA732981 MANA732982 MANA732983 MANA732984 MANA732985 MANA732986 MANA732987
Flash chomatography cartridges, empty colums, CHROMABOND® Flash DL and connecting kits
Kromatografiakolonnit
CHROMABOND® Flash DL cartridges (empty columns) can be used to pack individual adsorbents.
  • Heavy-duty polypropylene cartridges for self-filling
  • Can be used as solid injection system
  • Female Luer lock inlet / male Luer lock outlet
  • Suitable connecting kits
For solid injection a solid sample is dissolved in a suitable solvent and adsorbed onto CHROMABOND® XTR (diatomaceous earth). After removal of the residual solvent, the adsorbent is put into an empty CHROMABOND® Flash DL cartridge.
The self-filled cartridges can be connected to a flash instrumentMore Product Information