SPE columns, drugs of abuse analysis, CHROMABOND® Drug
Toimittaja: MACHEREY-NAGEL
730168 732168 738161.100M 730168.250 730681
MANA730168EA 377 EUR
MANA730168 MANA732168 MANA738161.100M MANA730168.250 MANA730681
SPE columns, drugs of abuse analysis, CHROMABOND® Drug
Kromatografiakolonnit
CHROMABOND® Drug is a special silica phase for the SPE enrichment of drugs of abuse from urine or plasma.
  • Base material: Silica
  • Special bifunctional modification: C8 / SA (strong cation exchanger)
Application: Enrichment of acidic, neutral and basic drugs from urine or plasma.