SPE columns, CHROMABOND® PSA
Toimittaja: MACHEREY-NAGEL
730462 730464
MANA730462EA 427 EUR
MANA730462 MANA730464
SPE columns, CHROMABOND® PSA
Kromatografiakolonnit
CHROMABOND® PSA is a propylsulfonic acid modified silica cation exchanger for SPE.
  • Base material: Silica
  • Very strong cation exchanger
  • Contrary to SCX phase (e.g. CHROMABOND® SA) no π-π interactions
Recommended for weak cations like cationic alkaloids.
Cation exchange capacity: ~0,7 meq/g