Empty solid phase extraction columns, CHROMABOND®
Toimittaja: MACHEREY-NAGEL
730103 730158 730159 730160 730161 730171 730172 730230 730355 730380 730382 730389 730474 732501
MANA730103EA 222 EUR
MANA730103 MANA730158 MANA730159 MANA730160 MANA730161 MANA730171 MANA730172 MANA730230 MANA730355 MANA730380 MANA730382 MANA730389 MANA730474 MANA732501
Empty solid phase extraction columns, CHROMABOND®
Kromatografiakolonnit
Empty and reservoir columns with accessories for solid phase extraction.
  • Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
  • Reservoir columns from PP for application of medium-size samples
  • Accessories for SPE columns (filter elements, adapters)