Capillary columns, OPTIMA® 1701 MS
Toimittaja: MACHEREY-NAGEL
726633.60 726630.30 726631.60 726630.60 726631.30 726633.30 726634.30 726634.60
MANA726633.60EA 2600 EUR
MANA726633.60 MANA726630.30 MANA726631.60 MANA726630.60 MANA726631.30 MANA726633.30 MANA726634.30 MANA726634.60
Capillary columns, OPTIMA® 1701 MS
Kromatografiakolonnit
Medium polar silarylene GC phase with low bleeding.
  • Ideal for ion trap and quadrupol MS detectors
  • Suitable for environmental analysis such as PAH, PCB and pesticides, also as reference column in combination with 5 MS columns
  • Maximum temperature for isothermal operation 280 °C
  • Maximum temperature for short isotherms in a temperature program 300 °C
The selectivity is analogue to 14% cyanopropyl-phenyl - 86% dimethylpolysiloxane phases.
USP code G46