Accessories for HPLC columns
Toimittaja: MACHEREY-NAGEL
706290 718582 718583 718584 718637 718755 718760 718761 718762 718763 718765 718766 718767 718768 718769 718770 718771 718772 718775 719489 718778
MANA706290EA 157 EUR
MANA706290 MANA718582 MANA718583 MANA718584 MANA718637 MANA718755 MANA718760 MANA718761 MANA718762 MANA718763 MANA718765 MANA718766 MANA718767 MANA718768 MANA718769 MANA718770 MANA718771 MANA718772 MANA718775 MANA719489 MANA718778
Accessories for HPLC columns
Kromatografiakolonnit
Accessories for stainless steel columns, PEEK fittings and capillaries.
  • Corrosion resistant
  • Pressure stable
Meets all requirements of HPLC applications.