Bulk media for HPLC, POLYGOPREP
Toimittaja: MACHEREY-NAGEL
711001.1000 711002.1000 711003.1000 711004.1000 711005.1000 711006.1000 711007.100 711007.1000 711008.100 711008.1000 711009.100 711009.1000 711011.100 711011.1000 711012.100 711012.1000 711013.100 711013.1000 711014.100 711014.1000 711015.100 711015.1000 711016.100 711016.1000 711017.100 711017.1000 711018.100 711018.1000 711019.100 711019.1000 711021.100 711021.1000 711022.100 711022.1000 711023.100 711023.1000 711024.100 711024.1000 711025.100 711025.1000 711026.100 711026.1000 711027.100 711027.1000 711028.100 711028.1000 711029.100 711029.1000 711031.100 711031.1000 711032.100 711032.1000 711033.1000 711034.1000 711035.1000 711036.1000 711037.1000 711240.1000 711250.1000 711260.1000 711270.1000 711470.100 711470.1000 711480.100 711480.1000 711490.100 711490.1000 711500.100 711500.1000 711540.1000 711550.1000 711590.100 711590.1000 711610.1000 711620.1000 711630.1000 711690.100 711690.1000 711700.100 711700.1000 711720.100 711720.1000 711730.100 711730.1000
MANA711001.1000EA 688 EUR
MANA711001.1000 MANA711002.1000 MANA711003.1000 MANA711004.1000 MANA711005.1000 MANA711006.1000 MANA711007.100 MANA711007.1000 MANA711008.100 MANA711008.1000 MANA711009.100 MANA711009.1000 MANA711011.100 MANA711011.1000 MANA711012.100 MANA711012.1000 MANA711013.100 MANA711013.1000 MANA711014.100 MANA711014.1000 MANA711015.100 MANA711015.1000 MANA711016.100 MANA711016.1000 MANA711017.100 MANA711017.1000 MANA711018.100 MANA711018.1000 MANA711019.100 MANA711019.1000 MANA711021.100 MANA711021.1000 MANA711022.100 MANA711022.1000 MANA711023.100 MANA711023.1000 MANA711024.100 MANA711024.1000 MANA711025.100 MANA711025.1000 MANA711026.100 MANA711026.1000 MANA711027.100 MANA711027.1000 MANA711028.100 MANA711028.1000 MANA711029.100 MANA711029.1000 MANA711031.100 MANA711031.1000 MANA711032.100 MANA711032.1000 MANA711033.1000 MANA711034.1000 MANA711035.1000 MANA711036.1000 MANA711037.1000 MANA711240.1000 MANA711250.1000 MANA711260.1000 MANA711270.1000 MANA711470.100 MANA711470.1000 MANA711480.100 MANA711480.1000 MANA711490.100 MANA711490.1000 MANA711500.100 MANA711500.1000 MANA711540.1000 MANA711550.1000 MANA711590.100 MANA711590.1000 MANA711610.1000 MANA711620.1000 MANA711630.1000 MANA711690.100 MANA711690.1000 MANA711700.100 MANA711700.1000 MANA711720.100 MANA711720.1000 MANA711730.100 MANA711730.1000
Bulk media for HPLC, POLYGOPREP
HPLC- ja TLC, Täytemateriaalit Kromatografiakolonnien täytemateriaalit
Irregular silica phase for preparative HPLC.
  • Range of pore sizes 60, 100, 300 and 1000 Å
  • Available mean particle sizes 12 µm, 20 µm, 30 µm, 50 µm, 80 µm and 130 µm
  • Different modifications
  • pH stability 2 to 8
The irregular silica POLYGOPREP can be used for different preperative applications.