Bulk media for HPLC, POLYGOPREP
Toimittaja: MACHEREY-NAGEL
711001.1000
711002.1000
711003.1000
711004.1000
711005.1000
711006.1000
711007.100
711007.1000
711008.100
711008.1000
711009.100
711009.1000
711011.100
711011.1000
711012.100
711012.1000
711013.100
711013.1000
711014.100
711014.1000
711015.100
711015.1000
711016.100
711016.1000
711017.100
711017.1000
711018.100
711018.1000
711019.100
711019.1000
711021.100
711021.1000
711022.100
711022.1000
711023.100
711023.1000
711024.100
711024.1000
711025.100
711025.1000
711026.100
711026.1000
711027.100
711027.1000
711028.100
711028.1000
711029.100
711029.1000
711031.100
711031.1000
711032.100
711032.1000
711033.1000
711034.1000
711035.1000
711036.1000
711037.1000
711240.1000
711250.1000
711260.1000
711270.1000
711470.100
711470.1000
711480.100
711480.1000
711490.100
711490.1000
711500.100
711500.1000
711540.1000
711550.1000
711590.100
711590.1000
711610.1000
711620.1000
711630.1000
711690.100
711690.1000
711700.100
711700.1000
711720.100
711720.1000
711730.100
711730.1000
MANA711001.1000EA
688
EUR
MANA711001.1000
MANA711002.1000
MANA711003.1000
MANA711004.1000
MANA711005.1000
MANA711006.1000
MANA711007.100
MANA711007.1000
MANA711008.100
MANA711008.1000
MANA711009.100
MANA711009.1000
MANA711011.100
MANA711011.1000
MANA711012.100
MANA711012.1000
MANA711013.100
MANA711013.1000
MANA711014.100
MANA711014.1000
MANA711015.100
MANA711015.1000
MANA711016.100
MANA711016.1000
MANA711017.100
MANA711017.1000
MANA711018.100
MANA711018.1000
MANA711019.100
MANA711019.1000
MANA711021.100
MANA711021.1000
MANA711022.100
MANA711022.1000
MANA711023.100
MANA711023.1000
MANA711024.100
MANA711024.1000
MANA711025.100
MANA711025.1000
MANA711026.100
MANA711026.1000
MANA711027.100
MANA711027.1000
MANA711028.100
MANA711028.1000
MANA711029.100
MANA711029.1000
MANA711031.100
MANA711031.1000
MANA711032.100
MANA711032.1000
MANA711033.1000
MANA711034.1000
MANA711035.1000
MANA711036.1000
MANA711037.1000
MANA711240.1000
MANA711250.1000
MANA711260.1000
MANA711270.1000
MANA711470.100
MANA711470.1000
MANA711480.100
MANA711480.1000
MANA711490.100
MANA711490.1000
MANA711500.100
MANA711500.1000
MANA711540.1000
MANA711550.1000
MANA711590.100
MANA711590.1000
MANA711610.1000
MANA711620.1000
MANA711630.1000
MANA711690.100
MANA711690.1000
MANA711700.100
MANA711700.1000
MANA711720.100
MANA711720.1000
MANA711730.100
MANA711730.1000
Bulk media for HPLC, POLYGOPREP
HPLC- ja TLC, Täytemateriaalit
Kromatografiakolonnien täytemateriaalit
Irregular silica phase for preparative HPLC.
- Range of pore sizes 60, 100, 300 and 1000 Å
- Available mean particle sizes 12 µm, 20 µm, 30 µm, 50 µm, 80 µm and 130 µm
- Different modifications
- pH stability 2 to 8
The irregular silica POLYGOPREP can be used for different preperative applications.