Extraction thimbles, borosilicate glass
Toimittaja: MACHEREY-NAGEL
649111 649103 649104 649106 649107 649108 649109 649114 649118 649121 649122 649123 649124 649126 649128 649131 649135 649138 649143 649500 649501 649120
MANA649111EA 355 EUR
MANA649111 MANA649103 MANA649104 MANA649106 MANA649107 MANA649108 MANA649109 MANA649114 MANA649118 MANA649121 MANA649122 MANA649123 MANA649124 MANA649126 MANA649128 MANA649131 MANA649135 MANA649138 MANA649143 MANA649500 MANA649501 MANA649120
Extraction thimbles, borosilicate glass
Uuttolaitteet ja uutoshylsyt Uutoshylsyt
Made from glass micro fibers without binder.
  • Pollution monitoring, air sampling
  • High temperature filtration (up to 500 °C)
  • For solvents incompatible with cellulose thimbles (for example strong acids or bases)
Extraction thimbles for high temperatures and aggressive solvents