Phase separating paper
Toimittaja: MACHEREY-NAGEL
484011 484015 584011 584015 584012 430009 430011 430012 430015 430018 430024 484005 484007 484009 484012 484018 484024 484032 584009 584018 584024 584032
MANA484011EA 45.9 EUR
MANA484011 MANA484015 MANA584011 516-6012 516-6019 MANA430009 MANA430011 MANA430012 MANA430015 MANA430018 MANA430024 MANA484005 MANA484007 MANA484009 MANA484012 MANA484018 MANA484024 MANA484032 MANA584009 MANA584018 MANA584024 MANA584032
Phase separating paper
Suodattimet Suodatinpaperit Faasierotus
These papers are made hydrophobic (impermeable to water) by impregnation with a silicone. With the aid of these filters, water can be separated from water-immiscible organic solvents in an elegant manner, by means of a simple filtration.