Glass microfiber filters without binder, grade MN GF-2
Toimittaja: MACHEREY-NAGEL
4120025 4120037 4120045 412005 412007 412009 412011 412012 412015 412018 412024 412027
MANA4120025EA 81 EUR
MANA4120025 MANA4120037 MANA4120045 MANA412005 MANA412007 MANA412009 MANA412011 MANA412012 MANA412015 MANA412018 MANA412024 MANA412027
Glass microfiber filters without binder, grade MN GF-2
Suodattimet Lasikuitusuodattimet
MN GF-2 is a binder-free glass fiber filter paper with three times higher basis weight than MN GF-1. It combines fine particle retention with excellent loading capacity.
  • High basis weight
  • Excellent loading capacity
  • Binder-free