Glass microfiber filters without binder, grade MN GF-1
Toimittaja: MACHEREY-NAGEL
411009 4110025 4110037 4110045 411005 411007 411011 411012 411015 411018 411024 411027
MANA411009EA 143 EUR
MANA411009 MANA4110025 MANA4110037 MANA4110045 MANA411005 MANA411007 MANA411011 MANA411012 MANA411015 MANA411018 MANA411024 MANA411027
Glass microfiber filters without binder, grade MN GF-1
Suodattimet Lasikuitusuodattimet
MN GF-1 is a binder-free glass fiber filter paper that offers fine particle retention, high flow rate and good loading capacity. It is used for high efficiency laboratory filtration as well as air sampling. It is manufactured from borosilicate glass fiber and is resistant towards most organic and inorganic solvents.
  • High flow rate
  • Good loading capacity
  • Binder-free