Quantitative filter papers, ashless grades, MN 640
Toimittaja: MACHEREY-NAGEL
201007 201009 202007 202009 202011 202012 202015 202018 203005 203007 203009 203011 203012 203015 204015 205009 205011 205012 205015 205018 206009 206011 206012 206015 207005 207012 205007 201005 201011 201012 201015 201018 201024 201032 202005 202024 202032 203018 203024 203032 204005 204007 204009 204011 204012 204018 204024 204032 205005 205024 205032 206005 206007 206018 206024 206032 207007 207009 207011 207015 207018 207024 207032
MANA201007EA 31.7 EUR
MANA201007 MANA201009 512-0486 515-9700 512-0487 515-9701 512-0488 MANA202018 MANA203005 512-0490 MANA203009 512-0491 515-9710 512-0492 MANA204015 512-0496 512-0497 515-9800 MANA205015 MANA205018 MANA206009 MANA206011 MANA206012 MANA206015 MANA207005 MANA207012 512-0495 MANA201005 MANA201011 MANA201012 MANA201015 MANA201018 MANA201024 MANA201032 512-0084 512-0489 MANA202032 512-0493 MANA203024 MANA203032 MANA204005 MANA204007 MANA204009 MANA204011 512-0494 MANA204018 MANA204024 MANA204032 MANA205005 MANA205024 MANA205032 MANA206005 MANA206007 MANA206018 MANA206024 MANA206032 MANA207007 MANA207009 MANA207011 MANA207015 MANA207018 MANA207024 MANA207032
Quantitative filter papers, ashless grades, MN 640
Suodattimet Suodatinpaperit Kvantitatiivinen suodatinpaperi
These quantitative filter papers are made from refined pulp and linters. They are purified in a highly developed clean-up process to remove organic and inorganic impurities. This ensures a low ash content of <0,01 %. They are consistent performance and give safe results in gravimetric analyses.
  • Very low ash content
  • Suitable for gravimetric, quantitative analysis
  • Contamination-free filtration
Grade MN 640 dd 'green ribbon' is an ashless, slow filter paper which is slightly less retentive than MN 640 d.
Grade MN 640 d 'blue ribbon' is an ashless, slowMore Product Information