2-Propanoli, AR®, Laboratory Reagent, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials

Synonyms: Isopropyylialkoholi

6775-25EA 65.77 EUR
6775-25 6775-45 6775-39 MACR6775-16
2-Propanoli, AR®, Laboratory Reagent, Macron Fine Chemicals™
2-Propanoli
Kaava: (CH₃)₂CHOH
MDL numero: MFCD00011674
CAS-numero: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR