Dextrose, anhydrous, granular USP, Macron Fine Chemicals™

Toimittaja: Avantor Performance Materials

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

MACR4908-06EA 185 EUR
MACR4908-06 MACR4908-20 MACR4908-88 MACR4908-24
Dextrose, anhydrous, granular USP, Macron Fine Chemicals™
D(+)-glukoosi
Kaava: C₆H₁₂O₆
MW: 180.16 g/mol
Sulamispiste: 146
MDL numero: MFCD00148912
CAS-numero: 50-99-7
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

USP - Acidity (ml) ≤0.30
USP - Arsenic (As) ≤1 ppm
USP - Chloride (Cl) ≤0.018%
USP - Color of Aqueous Solution Passes Test
USP - Dextrin Passes Test
USP - Endotoxin Concentration (EU/g) ≤2.5
USP - Heavy Metals (as Pb) ≤0.0005%
USP - Identification Passes Test
USP - Residue on Ignition ≤0.1%
USP - Solubility Passes Test
USP - Soluble Starch, Sulfites Passes Test
USP - Specific Rotation [+]²5^D (+) 52.6 - 53.2 Degree
USP - Sulphate (SO₄) ≤0.025%
USP - Water (H₂O) ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR