DNA/RNA isolation, viral DNA/RNA 96 kit, Mag-Bind®
Toimittaja: OMEGA BIO-TEK
M6246-01 M6246-02 M6246-03
M6246-01EA 349 EUR
M6246-01 M6246-02 M6246-03
DNA/RNA isolation, viral DNA/RNA 96 kit, Mag-Bind®
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
Mag-Bind® Viral DNA/RNA kit is designed for the rapid and reliable isolation of viral RNA and viral DNA from whole blood, serum, plasma, saliva and other body fluids. The Mag-Bind® magnetic beads technology enables purification of high quality nucleic acids that is free of proteins, nucleases, and other impurities. In addition to easily being adapted with automated systems, this procedure can also be scaled up or down, allowing for the purification from various amounts of starting materials. The purified nucleic acid is ready for direct use in downstream applications, such as amplification or other enzymatic reactions.
  • Rapid isolation of high quality, ready to use viral DNA and RNA
  • No phenol/chloroform extraction
  • Purification from various amounts of starting material
  • Reproducible DNA and RNA purification from a variety of samples