VWR®, Bradford protein assay, proteomics grade
Toimittaja: VWR Chemicals
M172-1LEA 306 EUR
M172-1L
VWR®, Bradford protein assay, proteomics grade
Bio-assayt Proteiinianalyysit
The Bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration. Based upon complex formation between basic and aromatic amino acid residues with Coomassie® Brilliant Blue G-250 dye, the Bradford method is easier, faster, and more sensitive than the Lowry method.
  • Sensitive, fast and consistent
  • Standard protocol for measuring concentrations from 10 to 100 μg
  • Microassay protocol for measuring concentrations from 1 to 10 μg
Toimitustiedot: Each kit includes Bradford Reagent (1 L) and bovine albumin standard, 0,5 mg/ml (1,5 ml). Bradford Reagent and BSA standard solution are also available separately.