Natriumtiosulfaatti-5 H2O ≥99%, Proteomics Grade

Toimittaja: VWR Chemicals

M132-500GEA 167 EUR
M132-500G
Natriumtiosulfaatti-5 H2O ≥99%, Proteomics Grade
Natriumtiosulfaatti-5 H2O
Kaava: Na₂S₂O₃.5H₂O
Sulamispiste: 49 °C
Tiheys: 1,67 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149186
CAS-numero: 10102-17-7
EINECS: 231-867-5
ADR: ,
Merck Index: 12,08844

Order Now

Testitulosten erittely

Insolubles <0.005%
pH (5%, Water) @25 °C 6 – 8.4
Protease none detected
Purity >99%
Sulphate and Sulphite <0.1%
Sulphide <0.0001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR