Mag-Bind® Plant DNA plus kit
Toimittaja: OMEGA BIO-TEK
M1128-00 M1128-01
M1128-00EA 174 EUR
M1128-00 M1128-01
Mag-Bind® Plant DNA plus kit
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
The Mag-Bind® Plant DNA 96 plus kit allows rapid and reliable isolation of high-quality genomic DNA from a wide variety of difficult to lyse and high polysaccharide plant species and tissues. The system utilises a CTAB-based lysis buffer without the need for a chloroform or other organic solvent steps. The proprietary wash buffer systems eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. This kit is designed for manual or fully automated high throughput preparation of genomic, chloroplast, and mitochondrial DNA. The Mag-Bind® Plant DNA plus kit also provides greater flexibility than silica plate based systems as partial plates can be used and can be scaled up to a 24-well format with the appropriate magnetic stand. The streamlined protocol can be easily adapted on most robotic liquid handling platforms. Purified DNA is suitable for PCR, qPCR, restriction digestion, hybridisation applications, as well as more demanding applications, such as RAPD.
  • Simple, rapid, and reliable procedure
  • Reproducible yields of pure, ready to use DNA
  • Adaptable to most robotic liquid handling platform