Stoppers, butyl, for diagnostic vials, WHEATON®
Toimittaja: DWK Life Sciences
600005
LSUK600005EA 152 EUR
LSUK600005
Stoppers, butyl, for diagnostic vials, WHEATON®
Tulpat Kumi
Stoppers are made from the pharmaceutical grade materials.
  • Suitable for freeze drying