Ympäristöanalytiikan pikatestit, Multiparameter portable photometers, Lovibond MD 200
Toimittaja: Lovibond
2860102 2882002 2892502 2860502 2862102 2889002 2889402 2861802 2860542 2862912 2861902
LOVI2860102EA 1140 EUR
LOVI2860102 LOVI2882002 LOVI2892502 LOVI2860502 LOVI2862102 LOVI2889002 LOVI2889402 LOVI2861802 LOVI2860542 LOVI2862912 LOVI2861902
Ympäristöanalytiikan pikatestit, Multiparameter portable photometers, Lovibond MD 200
Ympäristöanalytiikan pikatestit
The versatile, compact benchtop photometer MD 200 combines high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic. The result: precise measurement results at all times - no matter where you use it.
  • Versatile use
  • Precise and low maintenance
  • User friendly
The MD 200 photometer can be used in almost all areas of water analysis. The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance. The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe. Use long-term stable Lovibond® reagent tablets (5 to 10 years guaranteed minimum durability) or Lovibond® liquid reagents.
Toimitustiedot: Instrument supplied with carry case, 4× AA batteries, 3× round glass vials with lids, relevant reagents, and instruction manual.