Accessories for comparators
Toimittaja: Lovibond
232110 232510 234070 283620 230380 232020 232040 232050 232060 232080 232520 233200 233960 233990 234030 236710 236770 283310 283320 283430 283570 283790 283800 283810 283940 292160 283530 230690
LOVI232110EA 724 EUR
LOVI232110 LOVI232510 LOVI234070 LOVI283620 LOVI230380 LOVI232020 LOVI232040 LOVI232050 LOVI232060 LOVI232080 LOVI232520 LOVI233200 LOVI233960 LOVI233990 LOVI234030 LOVI236710 LOVI236770 LOVI283310 LOVI283320 LOVI283430 LOVI283570 LOVI283790 LOVI283800 LOVI283810 LOVI283940 LOVI292160 LOVI283530 LOVI230690
Accessories for comparators
Kolorimetrit Vertailupakkaukset
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+