Test kits, copper (kits and reagents), CHECKIT®
Toimittaja: Lovibond
530300 147430 147440 147431
LOVI530300EA 89.5 EUR
LOVI530300 LOVI147430 LOVI147440 LOVI147431
Test kits, copper (kits and reagents), CHECKIT®
Ympäristöanalytiikan pikatestit
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
Toimitustiedot: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.