LiChrospher® RP-8, HPLC Columns
Toimittaja: Avantor
LISPRP8-5-125AEA 881 EUR
LISPRP8-5-125A LISPRP8-5125AF LISPRP8-5-250A LISPRP8-5250AF LISPRP8-5-50A LISPRP8-5-10C5 LISPRP8510C5SK
LiChrospher® RP-8, HPLC Columns
Kromatografiakolonnit
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • High efficiency packed columns