LiChrosorb® RP-8, HPLC Columns
Toimittaja: Avantor
LIRP8-10-250AEA 961 EUR
LIRP8-10-250A LIRP8-5-125A LIRP8-5-150A LIRP8-5-250A LIRP8-10-250AF LIRP8-5-10C5 LIRP8-10-200A
LiChrosorb® RP-8, HPLC Columns
Kromatografiakolonnit
These columns with high purity base deactivated porous silica are ideal for the separation of basic molecules.
  • Irregular porous silica
  • High efficiency and complimentary phase selectivity