Legionella AB Supplement
Toimittaja: LIOFILCHEM
81099
LIOF81099EA 0 EUR
LIOF81099
Legionella AB Supplement
Elatusaineet mikrobiologiaan Elatusaineiden lisäaineet, mikrobiologia
Legionella AB supplement is a lyophilised selective supplement used in combination with legionella growth supplement for the preparation of legionella BCYE agar base. The complete medium is used for detection and enumeration of legionella spp from clinical specimens and environmental sources including all kinds of water samples.
Sertifikaatit: ISO 11731.
Toimitustiedot: Each kit contains ten vials of freeze-dried legionella AB supplement, one instruction sheet.