EnteroPluri test
Toimittaja: LIOFILCHEM
71618 80252 71640 80257 80260 78619 78618 80281 71709 72560 71708 78620 78621 87002
LIOF71618EA 208 EUR
LIOF71618 LIOF80252 LIOF71640 LIOF80257 LIOF80260 LIOF78619 LIOF78618 LIOF80281 LIOF71709 LIOF72560 LIOF71708 LIOF78620 LIOF78621 LIOF87002
EnteroPluri test
Pikatestit patogeenien ja allergeenien osoittamiseen Enzymatic Tests
EnteroPluri Test is a 12-sector system containing special culture media that permits identification of the Enterobacteriaceae and other gram negative, oxidase negative bacteria. The system allows the simultaneous inoculation of all media present in the sectors and the execution of 15 biochemical reactions. Microorganism is identified evaluating the colour change of the different culture media after 18 to 24 hours of incubation at 36 ±1 °C and by a code number obtained from biochemical reaction interpretation. Oxidase, Kovacs Reagent Indol (e.g. 1.00181.0002, 1.09293.0100, 1.11350.0001) and Codebook are not included.
  • Based on biochemical tests performed on culture media containing specific substrates
  • Combination of positive and negative permits identification of bacteria by using the Codebook