Dehydrated culture media
Toimittaja: LIOFILCHEM
610004 610014 610021 610024 610040 610113 610036 610106 620071 620194 621016 610095 621007 620610 610136 610154 620125 620134 610135 610187 620601 620611 610300 610241 610207 610186 611022 610341 610303 610048 610190 610309 620020 620090 620176 620613 610360 610192 610371 610094 620114 620095 610218 610195 610142 610111 610358 620239 620630 620349 620350 620108 620614 610369 610088 610378 620102 612501 610310 610610 610160 610205 622202 610389 610390 610108 610140 610150 610163 610233 610311 610005 610012 611010 610623 610627 610611 610031 610137 610307 610125 610614 620077 611265 610144 610385 610098 610387 610356 610054 610027 610093 610018 610617 610628 620617 610370 620205 610182 611214 610164
LIOF610004EA 210 EUR
LIOF610004 LIOF610014 LIOF610021 LIOF610024 LIOF610040 LIOF610113 LIOF610036 LIOF610106 LIOF620071 LIOF620194 LIOF621016 LIOF610095 LIOF621007 LIOF620610 LIOF610136 LIOF610154 LIOF620125 LIOF620134 LIOF610135 LIOF610187 LIOF620601 LIOF620611 LIOF610300 LIOF610241 LIOF610207 LIOF610186 LIOF611022 LIOF610341 LIOF610303 LIOF610048 LIOF610190 LIOF610309 LIOF620020 LIOF620090 LIOF620176 LIOF620613 LIOF610360 LIOF610192 LIOF610371 LIOF610094 LIOF620114 LIOF620095 LIOF610218 LIOF610195 LIOF610142 LIOF610111 LIOF610358 LIOF620239 LIOF620630 LIOF620349 LIOF620350 LIOF620108 LIOF620614 LIOF610369 LIOF610088 LIOF610378 LIOF620102 LIOF612501 LIOF610310 LIOF610610 LIOF610160 LIOF610205 LIOF622202 LIOF610389 LIOF610390 LIOF610108 LIOF610140 LIOF610150 LIOF610163 LIOF610233 LIOF610311 LIOF610005 LIOF610012 LIOF611010 LIOF610623 LIOF610627 LIOF610611 LIOF610031 LIOF610137 LIOF610307 LIOF610125 LIOF610614 LIOF620077 LIOF611265 LIOF610144 LIOF610385 LIOF610098 LIOF610387 LIOF610356 LIOF610054 LIOF610027 LIOF610093 LIOF610018 LIOF610617 LIOF610628 LIOF620617 LIOF610370 LIOF620205 LIOF610182 LIOF611214 LIOF610164
Dehydrated culture media
Elatusaineet mikrobiologiaan Elatusaineet mikrobiologiaan, jauhemaiset
Dehydrated culture media in powder available in 100 g, 500 g and 5 kg formats.
  • Produced in a strictly controlled environment according to the highest quality standards
  • ISO 9001 and ISO 13485 certified manufacturing facility
  • The lid contains an induction seal to ensure the humidity control
  • Liofilchem produces over 3000 standard formulations in compliance with international standards including ISO, CLSI, EUCAST, APHA, AOAC, EP and USP
  • Flexibility for customising the medium composition