Dehydrated culture media
Toimittaja: LIOFILCHEM
610004
610014
610021
610024
610040
610113
610036
610106
620071
620194
621016
610095
621007
620610
610136
610154
620125
620134
610135
610187
620601
620611
610300
610241
610207
610186
611022
610341
610303
610048
610190
610309
620020
620090
620176
620613
610360
610192
610371
610094
620114
620095
610218
610195
610142
610111
610358
620239
620630
620349
620350
620108
620614
610369
610088
610378
620102
612501
610310
610610
610160
610205
622202
610389
610390
610108
610140
610150
610163
610233
610311
610005
610012
611010
610623
610627
610611
610031
610137
610307
610125
610614
620077
611265
610144
610385
610098
610387
610356
610054
610027
610093
610018
610617
610628
620617
610370
620205
610182
611214
610164
LIOF610004EA
210
EUR
LIOF610004
LIOF610014
LIOF610021
LIOF610024
LIOF610040
LIOF610113
LIOF610036
LIOF610106
LIOF620071
LIOF620194
LIOF621016
LIOF610095
LIOF621007
LIOF620610
LIOF610136
LIOF610154
LIOF620125
LIOF620134
LIOF610135
LIOF610187
LIOF620601
LIOF620611
LIOF610300
LIOF610241
LIOF610207
LIOF610186
LIOF611022
LIOF610341
LIOF610303
LIOF610048
LIOF610190
LIOF610309
LIOF620020
LIOF620090
LIOF620176
LIOF620613
LIOF610360
LIOF610192
LIOF610371
LIOF610094
LIOF620114
LIOF620095
LIOF610218
LIOF610195
LIOF610142
LIOF610111
LIOF610358
LIOF620239
LIOF620630
LIOF620349
LIOF620350
LIOF620108
LIOF620614
LIOF610369
LIOF610088
LIOF610378
LIOF620102
LIOF612501
LIOF610310
LIOF610610
LIOF610160
LIOF610205
LIOF622202
LIOF610389
LIOF610390
LIOF610108
LIOF610140
LIOF610150
LIOF610163
LIOF610233
LIOF610311
LIOF610005
LIOF610012
LIOF611010
LIOF610623
LIOF610627
LIOF610611
LIOF610031
LIOF610137
LIOF610307
LIOF610125
LIOF610614
LIOF620077
LIOF611265
LIOF610144
LIOF610385
LIOF610098
LIOF610387
LIOF610356
LIOF610054
LIOF610027
LIOF610093
LIOF610018
LIOF610617
LIOF610628
LIOF620617
LIOF610370
LIOF620205
LIOF610182
LIOF611214
LIOF610164
Dehydrated culture media
Elatusaineet mikrobiologiaan
Elatusaineet mikrobiologiaan, jauhemaiset
Dehydrated culture media in powder available in 100 g, 500 g and 5 kg formats.
- Produced in a strictly controlled environment according to the highest quality standards
- ISO 9001 and ISO 13485 certified manufacturing facility
- The lid contains an induction seal to ensure the humidity control
- Liofilchem produces over 3000 standard formulations in compliance with international standards including ISO, CLSI, EUCAST, APHA, AOAC, EP and USP
- Flexibility for customising the medium composition