Contact Plates, Blister
Toimittaja: LIOFILCHEM
15325 15327 15328 15329 15331 15332 15339S 15362 15334 15365 15323S 15326 15327S 15336 15340S 15375 15376 15380S 15360 15205 15336S 15361
LIOF15325EA 63.5 EUR
LIOF15325 LIOF15327 LIOF15328 LIOF15329 LIOF15331 LIOF15332 LIOF15339S LIOF15362 LIOF15334 LIOF15365 LIOF15323S LIOF15326 LIOF15327S LIOF15336 LIOF15340S LIOF15375 LIOF15376 LIOF15380S LIOF15360 LIOF15205 LIOF15336S LIOF15361
Contact Plates, Blister
Elatusaineet mikrobiologiaan Kontaktimaljat mikrobiologiaan
The peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and in controlled environment, offers the following advantages compared to the pouch pack:
  • Full transparency of the package
  • No release particles during the film opening phase, critical in environment controlled by particle counters
  • Allows the usage of single plates, leaving the remaining plates protected in their pack
  • Drastically reduces the dehydration of culture medium during the shelf life period
  • Permits better product preservation and isolation during transportation and storage
  • Guarantees the product cannot be handled after the packaging and avoids contamination