Ceramic beads, SiLibeads®
Toimittaja: SIGMUND LINDNER
9719 9707 9709 9711 9717 9729 9730 97025
LINR9719EA 557 EUR
LINR9719 LINR9707 LINR9709 LINR9711 LINR9717 LINR9729 LINR9730 LINR97025
Ceramic beads, SiLibeads®
Helmet ja partikkelit Ceramic Beads
Ceramic beads made of Zirconium Oxide / Yttrium stabilised.
  • High density
  • Very high wear and tear resistance
  • Low contamination of the grinding product
  • Useable for all modern types of mills and high energy mills (vertical and horizontal)
  • Excellent crystal structure avoids bead breakage and reduces the abrasion of mill parts
  • No radioactivity
  • Conformity to 1935/2004/EC
The exact roundness as well as the smooth and polished surface of the beads contributes to low wear rate.