Human IgA purification kit, LigaTrap®
Toimittaja: LIGATRAP TECHNOLOGIES
LT-146 LT-146KIT LT-146-1X1ML LT-146-1X5ML LT-146-3X1ML
LIGALT-146EA 0 EUR
LIGALT-146 LIGALT-146KIT LIGALT-146-1X1ML LIGALT-146-1X5ML LIGALT-146-3X1ML
Human IgA purification kit, LigaTrap®
Proteiininpuhdistusjärjestelmät Protein Purification Kits
LigaTrap® Human IgA Purification Kit with LT technology utilises novel, patented ligands, designed for IgA antibody purification. Kit provides all the necessary reagents for fast, convenient micro-scale purification of human IgA in just ten easy steps.
  • Designed to purify human IgA
  • For in vitro laboratory use
  • Binding capacity for monoclonal human IgA is >15 mg/ml resin
  • Storage: 2 to 8 °C
LigaTrap® Human IgA purification resin is engineered to purify high quality human IgA antibodies from recombinant and monoclonal sources. Resins bind an exceptionally broad range of recombinant, monoclonal and human/animal source polyclonal immunoglobulins.
Each spin column can be used, regenerated, and used up to ten times with minimal
loss in binding capacity.
Toimitustiedot: Each kit contains ten microspin columns prefilled with 0,10 ml of LigaTrap® human IgA purification resin, buffers and collection tubes.
Varoitus: Not for diagnostic or therapeutic use. Not for human or animal consumption.