Biology prepared microscope slides, school set D
Toimittaja: LIEDER JOHANNES
753F 762E 766F 770F 772D 775G 777F 778H 782D 784E 785E 786E 787E 789E 790E 791F 750 751C 752D 758D 761D 764D 765F 788E 800C 763F 759D 781F 794E 796C
LIED753FEA 24.8 EUR
LIED753F LIED762E LIED766F LIED770F LIED772D LIED775G LIED777F LIED778H LIED782D LIED784E LIED785E LIED786E LIED787E LIED789E LIED790E LIED791F 761-0102 LIED751C LIED752D LIED758D LIED761D LIED764D LIED765F LIED788E LIED800C LIED763F LIED759D LIED781F LIED794E LIED796C
Biology prepared microscope slides, school set D
Alus- ja peitinlasit Valmisdiat
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The sets contain slides of typical micro-organisms, of cell division and of embryonic developments, as well as of tissues and organs of plants, animals and man.
Toimitustiedot: Supplied on high quality ground edge slides.