Biology prepared microscope slides
Toimittaja: LIEDER JOHANNES
706D 707C 716E 717F 718D 719D 721C 722D 723D 726D 728D 729D 734B 735D 737D 738D 741D 746D 747D 700 701F 702F 703D 725D 727E 731C 732D 736E 740D 744D 745C 748B 708E 724E 749F 714D 7450
LIED706DEA 17.8 EUR
LIED706D LIED707C LIED716E LIED717F LIED718D LIED719D LIED721C LIED722D LIED723D LIED726D LIED728D LIED729D LIED734B LIED735D LIED737D LIED738D LIED741D LIED746D LIED747D LIED700 LIED701F LIED702F LIED703D LIED725D LIED727E LIED731C LIED732D LIED736E LIED740D LIED744D LIED745C LIED748B LIED708E LIED724E LIED749F LIED714D LIED7450
Biology prepared microscope slides
Alus- ja peitinlasit Valmisdiat
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
Toimitustiedot: Supplied on high quality ground edge slides (26×76 mm).