Biology prepared microscope slides, school set A
Toimittaja: LIEDER JOHANNES
508C 509D 511D 514C 519C 520C 522D 523D 525D 500 501E 502E 503C 504C 505D 507C 510D 512C 513D 515C 516C 517C 518C 521C 524C
LIED508CEA 17.2 EUR
LIED508C LIED509D LIED511D LIED514C LIED519C LIED520C LIED522D LIED523D LIED525D LIED500 LIED501E LIED502E LIED503C LIED504C LIED505D LIED507C LIED510D LIED512C LIED513D LIED515C LIED516C LIED517C LIED518C LIED521C LIED524C
Biology prepared microscope slides, school set A
Alus- ja peitinlasit Valmisdiat
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
Toimitustiedot: Supplied on high quality ground edge slides (26×76 mm).