Spark-free Performance laboratory refrigerators with ATEX certificate, SRFfg
Toimittaja: Liebherr
SRFFG4001
LIEBSRFFG4001EA 2155 EUR
LIEBSRFFG4001
Spark-free Performance laboratory refrigerators with ATEX certificate, SRFfg
Jääkaapit
The refrigerators with spark-free interior are easy to clean and the robust glass shelves are simple to remove at 90° door opening. They are super-efficient, utilising the energy-savings potential of every component.
  • Alarm relay, alarm test function and custom alarm settings possible
  • Min./max. temperature log, self-closing door at <90° and mechanical lock provided
  • Retrofittable optional LAN/WiFi, Local API (optional via SmartModule)
  • SmartModule can be retrofitted
  • Antimicrobial handle with opening mechanism
  • Super-efficient energy consumption and forced-