Under Counter Refrigerator, Rd 1401 Pure
Toimittaja: Liebherr
RD1401
LIEBRD1401EA 0 EUR
LIEBRD1401
Under Counter Refrigerator, Rd 1401 Pure
Jääkaapit
Rd 1401 Pure Under counter refrigerator with lever handle, provides SuperCool functionality that maintains the optimum temperature. Even if you have just put shopping in from the market which has been warmed by the sun, once SuperCool functionality is activated, Liebherr boosts its performance until the fresh food is as cool as the rest of the contents. SuperCool then switches off automatically.
  • Interior minimal UI control unit
  • Replaceable door seal
  • One bottle rack, two can racks and egg tray (10 eggs)
  • SuperCool technology, display lock, door alarm (