Mikrosuodatinsuppilot
Toimittaja: Lenz Laborglas GmbH & CO.KG
07190410 07190310 07190010 07190110 07190210
LENZ07190410EA 37.2 EUR
LENZ07190410 LENZ07190310 LENZ07190010 LENZ07190110 LENZ07190210
Mikrosuodatinsuppilot
Suppilot Suodatussuppilot Mikrosuodatussuppilot
Filter funnels, made of borosilicate glass 3,3, with integrated sintered glass filter plate.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance