Flange clamps
Toimittaja: Lenz Laborglas GmbH & CO.KG
06248515 06248525 06248550
201-0120EA 104 EUR
201-0120 LENZ06248525 LENZ06248550
Flange clamps
Kourat Yleiskäyttöiset puristimet
Plastic, black.
  • Easy to assemble and disassemble
  • Tough construction, good temperature resistance
  • Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.