Reaction vessels, cylindrical, with flat flange
Toimittaja: Lenz Laborglas GmbH & CO.KG
06061137-V2 06061149-V2 06061158-V2 06061249-V2 06061258-V2 06061270-V2 06061273-V2 06061276-V2
LENZ06061137-V2EA 2040 EUR
LENZ06061137-V2 LENZ06061149-V2 LENZ06061158-V2 LENZ06061249-V2 LENZ06061258-V2 LENZ06061270-V2 LENZ06061273-V2 LENZ06061276-V2
Reaction vessels, cylindrical, with flat flange
Reaktioastiat Reaktioastiat, sylinterin muotoiset
DURAN® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With thermostatic and vaccum jacket and withdrawal valve.
Valve bore of 10 mm