Reaction vessels, round bottom, with flat flange
Toimittaja: Lenz Laborglas GmbH & CO.KG
06020158 06020149 06020170 06020273 06020279 06020285 06020485 06020488 06021149 06021158 06021170 06021273 06021279 06021285 06021585 06021588
LENZ06020158EA 1720 EUR
LENZ06020158 LENZ06020149 LENZ06020170 LENZ06020273 LENZ06020279 LENZ06020285 LENZ06020485 LENZ06020488 LENZ06021149 LENZ06021158 LENZ06021170 LENZ06021273 LENZ06021279 LENZ06021285 LENZ06021585 LENZ06021588
Reaction vessels, round bottom, with flat flange
Reaktioastiat Reaktioastiat, pullomalli
DURAN® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Spherical, with thermostatic jacket and flange with groove.