Putket, koeputket NS-tulpalla
Toimittaja: Lenz Laborglas GmbH & CO.KG
05700207 05700210 05700214 05700226 05700230 05700234 05700238 05700242
LENZ05700207EA 9.5 EUR
LENZ05700207 LENZ05700210 201-1165 LENZ05700226 LENZ05700230 LENZ05700234 LENZ05700238 LENZ05700242
Putket, koeputket NS-tulpalla
Putket Koeputket
DURAN®, borosilicate glass, clear. With standard ground joint and hexagonal glass stopper.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes